ContactUsLinkComponent
Mouse Monoamine Oxidase A/MAO-A ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantit ative determination of mouse Monoamine Oxidase A/MAO-A in serum, plasma, tissue homogenates or other biological fluid 1 * 48 Tests
Catalog # ANTIA270667-48
Supplier:  ANTIBODIES.COM
New Product
Mouse Monoamine Oxidase A/MAO-A ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantit ative determination of mouse Monoamine Oxidase A/MAO-A in serum, plasma, tissue homogenates or other biological fluid 1 * 48 Tests
Catalog # ANTIA270667-48
Supplier:  ANTIBODIES.COM