ContactUsLinkComponent
Human Bcl-2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Bcl-2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA1842-48
Supplier:  Antibodies.com
New Product
Human Bcl-2 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human Bcl-2 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA1842-48
Supplier:  Antibodies.com