ContactUsLinkComponent
Goat IL-17A Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibod y, Streptavidin-AP, recombinant bovine IL-17A ELISA standard, and standard reconstitution buffer. This matched antibo 1 * 576 Tests
Catalog # ANTIA270384-696
Supplier:  Antibodies.com
New Product
Goat IL-17A Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibod y, Streptavidin-AP, recombinant bovine IL-17A ELISA standard, and standard reconstitution buffer. This matched antibo 1 * 576 Tests
Catalog # ANTIA270384-696
Supplier:  Antibodies.com