ContactUsLinkComponent
Human IL-12/IL-23 p40 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detecti on antibody, Streptavidin-HRP, recombinant human IL-12 p70 ELISA standard, and standard reconstitution buffer. This m 1 * 1.920 Tests
Catalog # ANTIA270485-2096
Supplier:  ANTIBODIES.COM
New Product
Human IL-12/IL-23 p40 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detecti on antibody, Streptavidin-HRP, recombinant human IL-12 p70 ELISA standard, and standard reconstitution buffer. This m 1 * 1.920 Tests
Catalog # ANTIA270485-2096
Supplier:  ANTIBODIES.COM