ContactUsLinkComponent
Mouse nNOS (neuronal) ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative dete rmination of mouse nNOS (neuronal) in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA270629-48
Supplier:  Antibodies.com
New Product
Mouse nNOS (neuronal) ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative dete rmination of mouse nNOS (neuronal) in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA270629-48
Supplier:  Antibodies.com