ContactUsLinkComponent
Human LPO ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human LPO in breast milk, tears or other biological fluids. 1 * 48 Tests
Catalog # ANTIA2400-48
Supplier:  Antibodies.com
New Product
Human LPO ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human LPO in breast milk, tears or other biological fluids. 1 * 48 Tests
Catalog # ANTIA2400-48
Supplier:  Antibodies.com