ContactUsLinkComponent
Human Lipoprotein A ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determ ination of human Lipoprotein A in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA1896-48
Supplier:  Antibodies.com
New Product
Human Lipoprotein A ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determ ination of human Lipoprotein A in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA1896-48
Supplier:  Antibodies.com