ContactUsLinkComponent
APC belongs to the phycobiliprotein family of highly soluble and fluorescent proteins derived from cyanobacteria and eukaryotic alga e. APC is an ultra-sensitive fluorescent tracer with high emission quantum yields, that emits in the near-infrared fl 1 * 10 mg
Catalog # ANTIA269984-10
Supplier:  ANTIBODIES.COM
New Product
APC belongs to the phycobiliprotein family of highly soluble and fluorescent proteins derived from cyanobacteria and eukaryotic alga e. APC is an ultra-sensitive fluorescent tracer with high emission quantum yields, that emits in the near-infrared fl 1 * 10 mg
Catalog # ANTIA269984-10
Supplier:  ANTIBODIES.COM