ContactUsLinkComponent
Hamster Interferon gamma Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal dete ction antibody, Streptavidin-AP, recombinant hamster Interferon gamma ELISA standard, and standard reconstitution buf 1 * 576 Tests
Hamster Interferon gamma Matched Antibody Pair Kit
Catalog # ANTIA270371-696
Supplier:  Antibodies.com
New Product
CAS Number:  
Hamster Interferon gamma Matched Antibody Pair Kit
Catalog # ANTIA270371-696
Supplier:  Antibodies.com
New Product
CAS Number:  
About this item
Frequently Bought Together