ContactUsLinkComponent
Human IRAKM ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human IRAKM in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA2600-48
Supplier:  Antibodies.com
New Product
Human IRAKM ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human IRAKM in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA2600-48
Supplier:  Antibodies.com