ContactUsLinkComponent
Human CCL4/MIP-1 beta ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative dete rmination of human CCL4/MIP-1 beta in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or oth 1 * 96 Tests
Catalog # ANTIA2806-96
Supplier:  ANTIBODIES.COM
New Product
Human CCL4/MIP-1 beta ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative dete rmination of human CCL4/MIP-1 beta in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or oth 1 * 96 Tests
Catalog # ANTIA2806-96
Supplier:  ANTIBODIES.COM