ContactUsLinkComponent
Rabbit IgG Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibody , Streptavidin-HRP, Rabbit IgG ELISA standard, and standard reconstitution buffer. This matched antibody pair kit can 1 * 576 Tests
Catalog # ANTIA270536-696
Supplier:  Antibodies.com
New Product
Rabbit IgG Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibody , Streptavidin-HRP, Rabbit IgG ELISA standard, and standard reconstitution buffer. This matched antibody pair kit can 1 * 576 Tests
Catalog # ANTIA270536-696
Supplier:  Antibodies.com