Zadání objednávky
Czech Republic
ContactUsLinkComponent
[EN]HUMAN CDKN2A ELISA KIT 1 * 48 Tests
Human CDKN2A ELISA Kit
Catalog # ANTIA1766-48
Supplier:  Antibodies.com
New Product
CAS Number:  
Human CDKN2A ELISA Kit
Catalog # ANTIA1766-48
Supplier:  Antibodies.com
New Product
CAS Number:  
About this item
Human CDKN2A ELISA kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human CDKN2A in tissue homogenates, cell lysates or other biological fluids.
Human CDKN2A ELISA Kit (A1766) employs the sandwich enzyme immunoassay technique for the quantitative measurement of human CDKN2A in tissue homogenates, cell lysates or other biological fluids. An antibody specific for CDKN2A has been pre-coated onto a 96-well microtiter plate. The standards and test samples are added into the wells and the CDKN2A present in each sample is bound to the wells by the immobilised antibody. Following incubation, the wells are washed and then incubated with Biotinylated Anti-CDKN2A Antibody, which binds the captured CDKN2A present in each well. Following incubation, unbound biotinylated detection antibody is removed by washing, and an HRP-Avidin conjugate is added to the wells and the microtiter plate is incubated. Following incubation and washing, TMB substrate solution is then used to visualise the HRP enzymatic reaction by catalysis to produce a blue-coloured product that changes to yellow after addition of acidic stop solution. The density of yellow is proportional to the amount of CDKN2A captured in each well. The concentration of CDKN2A can then be calculated by reading the O.D. absorbance at 450 nm in a microplate reader and referring to the standard curve.
Caution: This product is for Research Use Only. It is not for use in diagnostic or therapeutic procedures.
Frequently Bought Together