ContactUsLinkComponent
Human Neuropeptide S ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative de termination of human Neuropeptide S in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA2024-48
Supplier:  Antibodies.com
New Product
Human Neuropeptide S ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative de termination of human Neuropeptide S in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA2024-48
Supplier:  Antibodies.com