Human Monoamine Oxidase A/MAO-A ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantit ative determination of human Monoamine Oxidase A/MAO-A in serum, plasma, tissue homogenates or other biological fluid 1 * 96 Tests
Catalog # ANTIA270740-96
Supplier: ANTIBODIES.COM