ContactUsLinkComponent
Human Epstein-Barr Virus Capsid Antigen IgM ELISA Kit is a trapping Enzyme-Linked Immunosorbent Assay designed for the in vitro qual itative determination of human Epstein-Barr Virus Capsid Antigen IgM in serum, plasma, tissue homogenates, and other 1 * 96 Tests
Catalog # ANTIA247221-96
Supplier:  ANTIBODIES.COM
New Product
Human Epstein-Barr Virus Capsid Antigen IgM ELISA Kit is a trapping Enzyme-Linked Immunosorbent Assay designed for the in vitro qual itative determination of human Epstein-Barr Virus Capsid Antigen IgM in serum, plasma, tissue homogenates, and other 1 * 96 Tests
Catalog # ANTIA247221-96
Supplier:  ANTIBODIES.COM