ContactUsLinkComponent
Human Mast Cell Tryptase ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative d etermination of human Mast Cell Tryptase in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA2052-48
Supplier:  ANTIBODIES.COM
New Product
Human Mast Cell Tryptase ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative d etermination of human Mast Cell Tryptase in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA2052-48
Supplier:  ANTIBODIES.COM