ContactUsLinkComponent
Activated R-Phycoerythrin (R-PE) is an intensely bright phycobiliprotein isolated from newly identified red algae species exclusive to the coastal region of Taiwan. It exhibits extremely bright red-orange fluorescence with high quantum yields, and ge 1 * 10 mg
Catalog # ANTIA269983-10
Supplier:  ANTIBODIES.COM
New Product
Activated R-Phycoerythrin (R-PE) is an intensely bright phycobiliprotein isolated from newly identified red algae species exclusive to the coastal region of Taiwan. It exhibits extremely bright red-orange fluorescence with high quantum yields, and ge 1 * 10 mg
Catalog # ANTIA269983-10
Supplier:  ANTIBODIES.COM