ContactUsLinkComponent
Human IL-21 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibod y, Streptavidin-HRP, recombinant human IL-21 ELISA standard, and standard reconstitution buffer. This matched antibod 1 * 576 Tests
Catalog # ANTIA270490-696
Supplier:  Antibodies.com
New Product
Human IL-21 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection antibod y, Streptavidin-HRP, recombinant human IL-21 ELISA standard, and standard reconstitution buffer. This matched antibod 1 * 576 Tests
Catalog # ANTIA270490-696
Supplier:  Antibodies.com