ContactUsLinkComponent
Human Aquaporin 4 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determin ation of human Aquaporin 4 in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA1776-48
Supplier:  ANTIBODIES.COM
New Product
Human Aquaporin 4 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determin ation of human Aquaporin 4 in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA1776-48
Supplier:  ANTIBODIES.COM