ContactUsLinkComponent
Feline Interferon gamma Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detec tion antibody, Streptavidin-HRP, recombinant cat Interferon gamma ELISA standard, and standard reconstitution buffer. 1 * 1.920 Tests
Catalog # ANTIA270467-2096
Supplier:  ANTIBODIES.COM
New Product
Feline Interferon gamma Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detec tion antibody, Streptavidin-HRP, recombinant cat Interferon gamma ELISA standard, and standard reconstitution buffer. 1 * 1.920 Tests
Catalog # ANTIA270467-2096
Supplier:  ANTIBODIES.COM