ContactUsLinkComponent
Mouse IgA Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, an AP-labelled monoclonal detection antibody, native Native mouse IgA ELISA standard, and standard reconstitution buffer. This matched antibody pair kit can be us 1 * 576 Tests
Catalog # ANTIA270432-696
Supplier:  Antibodies.com
New Product
Mouse IgA Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, an AP-labelled monoclonal detection antibody, native Native mouse IgA ELISA standard, and standard reconstitution buffer. This matched antibody pair kit can be us 1 * 576 Tests
Catalog # ANTIA270432-696
Supplier:  Antibodies.com