ContactUsLinkComponent
Rat Amylin/DAP ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determin ation of rat Amylin/DAP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biologic 1 * 48 Tests
Catalog # ANTIA1945-48
Supplier:  ANTIBODIES.COM
New Product
Rat Amylin/DAP ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determin ation of rat Amylin/DAP in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biologic 1 * 48 Tests
Catalog # ANTIA1945-48
Supplier:  ANTIBODIES.COM