ContactUsLinkComponent
Feline Interferon gamma Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detec tion antibody, Streptavidin-AP, recombinant cat Interferon gamma ELISA standard, and standard reconstitution buffer. 1 * 576 Tests
Catalog # ANTIA270381-696
Supplier:  ANTIBODIES.COM
New Product
Feline Interferon gamma Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detec tion antibody, Streptavidin-AP, recombinant cat Interferon gamma ELISA standard, and standard reconstitution buffer. 1 * 576 Tests
Catalog # ANTIA270381-696
Supplier:  ANTIBODIES.COM