ContactUsLinkComponent
Advanced Glycation End Products/AGE ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of Advanced Glycation End Products/AGE in serum, plasma, tissue homogenates or other biolog 1 * 48 Tests
Catalog # ANTIA2345-48
Supplier:  ANTIBODIES.COM
New Product
Advanced Glycation End Products/AGE ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitro quantitative determination of Advanced Glycation End Products/AGE in serum, plasma, tissue homogenates or other biolog 1 * 48 Tests
Catalog # ANTIA2345-48
Supplier:  ANTIBODIES.COM