ContactUsLinkComponent
Porcine KGF/FGF7 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determina tion of porcine KGF/FGF7 in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA2825-48
Supplier:  ANTIBODIES.COM
New Product
Porcine KGF/FGF7 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determina tion of porcine KGF/FGF7 in serum, plasma, tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA2825-48
Supplier:  ANTIBODIES.COM