ContactUsLinkComponent
Mouse 5HT2C Receptor ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative deter mination of mouse 5HT2C Receptor in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA270904-48
Supplier:  ANTIBODIES.COM
New Product
Mouse 5HT2C Receptor ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative deter mination of mouse 5HT2C Receptor in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA270904-48
Supplier:  ANTIBODIES.COM