ContactUsLinkComponent
Activated APC belongs to the phycobiliprotein family of highly soluble and fluorescent proteins derived from cyanobacteria and eukar yotic algae. APC is an ultra-sensitive fluorescent tracer with high emission quantum yields, that emits in the near-i 1 * 5 mg
  ANTIA269986-5
New Product
Activated APC belongs to the phycobiliprotein family of highly soluble and fluorescent proteins derived from cyanobacteria and eukar yotic algae. APC is an ultra-sensitive fluorescent tracer with high emission quantum yields, that emits in the near-i 1 * 5 mg
  ANTIA269986-5