ContactUsLinkComponent
Mouse TNF alpha Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection ant ibody, Streptavidin-AP, recombinant mouse TNF alpha ELISA standard, and standard reconstitution buffer. This matched 1 * 576 Tests
Catalog # ANTIA270448-696
Supplier:  ANTIBODIES.COM
New Product
Mouse TNF alpha Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection ant ibody, Streptavidin-AP, recombinant mouse TNF alpha ELISA standard, and standard reconstitution buffer. This matched 1 * 576 Tests
Catalog # ANTIA270448-696
Supplier:  ANTIBODIES.COM