ContactUsLinkComponent
Mouse IL-12/IL-23 p40 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detecti on antibody, Streptavidin-HRP, recombinant mouse IL-12 p70 ELISA standard, and standard reconstitution buffer. This m 1 * 576 Tests
Catalog # ANTIA270521-696
Supplier:  ANTIBODIES.COM
New Product
Mouse IL-12/IL-23 p40 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detecti on antibody, Streptavidin-HRP, recombinant mouse IL-12 p70 ELISA standard, and standard reconstitution buffer. This m 1 * 576 Tests
Catalog # ANTIA270521-696
Supplier:  ANTIBODIES.COM