ContactUsLinkComponent
Rat ZnT4 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat ZnT4 in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA270995-48
Supplier:  ANTIBODIES.COM
New Product
Rat ZnT4 ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of rat ZnT4 in tissue homogenates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA270995-48
Supplier:  ANTIBODIES.COM