ContactUsLinkComponent
R-Phycoerythrin (R-PE) is an intensely bright phycobiliprotein isolated from newly identified red algae species exclusive to the coa stal region of Taiwan. It exhibits extremely bright red-orange fluorescence with high quantum yields, and generates f 1 * 500 mg
Catalog # ANTIA269982-500
Supplier:  ANTIBODIES.COM
New Product
R-Phycoerythrin (R-PE) is an intensely bright phycobiliprotein isolated from newly identified red algae species exclusive to the coa stal region of Taiwan. It exhibits extremely bright red-orange fluorescence with high quantum yields, and generates f 1 * 500 mg
Catalog # ANTIA269982-500
Supplier:  ANTIBODIES.COM