ContactUsLinkComponent
Rat LDL Receptor ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determina tion of rat LDL Receptor in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA270951-48
Supplier:  ANTIBODIES.COM
New Product
Rat LDL Receptor ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determina tion of rat LDL Receptor in tissue homogenates, cell lysates or other biological fluids. 1 * 48 Tests
Catalog # ANTIA270951-48
Supplier:  ANTIBODIES.COM