ContactUsLinkComponent
Activated APC belongs to the phycobiliprotein family of highly soluble and fluorescent proteins derived from cyanobacteria and eukar yotic algae. APC is an ultra-sensitive fluorescent tracer with high emission quantum yields, that emits in the near-i 1 * 1 mg
Catalog # ANTIA269986-1
Supplier:  ANTIBODIES.COM
New Product
Activated APC belongs to the phycobiliprotein family of highly soluble and fluorescent proteins derived from cyanobacteria and eukar yotic algae. APC is an ultra-sensitive fluorescent tracer with high emission quantum yields, that emits in the near-i 1 * 1 mg
Catalog # ANTIA269986-1
Supplier:  ANTIBODIES.COM