ContactUsLinkComponent
Human IL-12 p70 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection ant ibody, Streptavidin-AP, recombinant human IL-12 p70 ELISA standard (lyophilized), and standard reconstitution buffer. 1 * 1.920 Tests
Catalog # ANTIA270402-2096
Supplier:  ANTIBODIES.COM
New Product
Human IL-12 p70 Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal detection ant ibody, Streptavidin-AP, recombinant human IL-12 p70 ELISA standard (lyophilized), and standard reconstitution buffer. 1 * 1.920 Tests
Catalog # ANTIA270402-2096
Supplier:  ANTIBODIES.COM