ContactUsLinkComponent
Full-length transmembrane human CCR2 protein expressed on the cell surface of Virus like particles. 1 * 50 µG
Catalog # ANTIA325311-50
Supplier:  Antibodies.com
New Product
Full-length transmembrane human CCR2 protein expressed on the cell surface of Virus like particles. 1 * 50 µG
Catalog # ANTIA325311-50
Supplier:  Antibodies.com