ContactUsLinkComponent
Mouse Complement C5a ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative deter mination of mouse Complement C5a in serum, plasma, and other biological fluids. This ELISA kit only requires 25&#0181 1 * 96 Tests
Catalog # ANTIA323211-96
Supplier:  Antibodies.com
New Product
Mouse Complement C5a ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative deter mination of mouse Complement C5a in serum, plasma, and other biological fluids. This ELISA kit only requires 25&#0181 1 * 96 Tests
Catalog # ANTIA323211-96
Supplier:  Antibodies.com