ContactUsLinkComponent
Monkey Anti-Polyethylene Glycol IgG ELISA Kit is a indirect Enzyme-Linked Immunosorbent Assay designed for the in vitro quantitative determination of monkey Anti-Polyethylene Glycol IgG in serum, plasma, cell culture supernatant, cell or tissue lysa 1 * 96 Tests
Catalog # ANTIA326387-96
Supplier:  ANTIBODIES.COM
New Product
Monkey Anti-Polyethylene Glycol IgG ELISA Kit is a indirect Enzyme-Linked Immunosorbent Assay designed for the in vitro quantitative determination of monkey Anti-Polyethylene Glycol IgG in serum, plasma, cell culture supernatant, cell or tissue lysa 1 * 96 Tests
Catalog # ANTIA326387-96
Supplier:  ANTIBODIES.COM