ContactUsLinkComponent
Synthetic nanodiscs offer a stable and biologically relevant environment that closely mimics cell membranes and enables full-length transmembrane human Niemann Pick C1 Like 1/NPC1L1 protein to be purified and analysed in vitro. 1 * 10 µG
Catalog # ANTIA318423-10
Supplier:  Antibodies.com
New Product
Synthetic nanodiscs offer a stable and biologically relevant environment that closely mimics cell membranes and enables full-length transmembrane human Niemann Pick C1 Like 1/NPC1L1 protein to be purified and analysed in vitro. 1 * 10 µG
Catalog # ANTIA318423-10
Supplier:  Antibodies.com