ContactUsLinkComponent
A heterodimer of IL-12A (Arg23-Ser219) with a C-terminal human Fc tag and IL-12B (Ile23-Ser328) with a C-terminal 6xHis tag. The hum an Fc tag consists of amino acids Glu99-Ala330. 1 * 100 µG
Catalog # ANTIA317711-100
Supplier:  Antibodies.com
New Product
A heterodimer of IL-12A (Arg23-Ser219) with a C-terminal human Fc tag and IL-12B (Ile23-Ser328) with a C-terminal 6xHis tag. The hum an Fc tag consists of amino acids Glu99-Ala330. 1 * 100 µG
Catalog # ANTIA317711-100
Supplier:  Antibodies.com