ContactUsLinkComponent
Mouse Ubiquitin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinat ion of mouse Ubiquitin in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA3095-48
Supplier:  Antibodies.com
New Product
Mouse Ubiquitin ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinat ion of mouse Ubiquitin in serum, plasma or other biological fluids. 1 * 48 Tests
Catalog # ANTIA3095-48
Supplier:  Antibodies.com