ContactUsLinkComponent
Rat IL-1 alpha ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinati on of rat IL-1 alpha in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological 1 * 48 Tests
Catalog # ANTIA2968-48
Supplier:  ANTIBODIES.COM
New Product
Rat IL-1 alpha ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determinati on of rat IL-1 alpha in serum, plasma, tissue homogenates, cell lysates, cell culture supernates or other biological 1 * 48 Tests
Catalog # ANTIA2968-48
Supplier:  ANTIBODIES.COM