ContactUsLinkComponent
Human IL-2 Receptor alpha Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal det ection antibody, Streptavidin-AP, recombinant human CD25 ELISA standard, and standard reconstitution buffer. This mat 1 * 576 Tests
Catalog # ANTIA325343-696
Supplier:  Antibodies.com
New Product
Human IL-2 Receptor alpha Matched Antibody Pair Kit includes an unlabeled monoclonal capture antibody, biotin-labeled monoclonal det ection antibody, Streptavidin-AP, recombinant human CD25 ELISA standard, and standard reconstitution buffer. This mat 1 * 576 Tests
Catalog # ANTIA325343-696
Supplier:  Antibodies.com