ContactUsLinkComponent
Synthetic nanodiscs offer a stable and biologically relevant environment that closely mimics cell membranes and enables full-length transmembrane human Glucose Transporter GLUT4 protein to be purified and analysed in vitro. 1 * 50 µG
Catalog # ANTIA318441-50
Supplier:  ANTIBODIES.COM
New Product
Synthetic nanodiscs offer a stable and biologically relevant environment that closely mimics cell membranes and enables full-length transmembrane human Glucose Transporter GLUT4 protein to be purified and analysed in vitro. 1 * 50 µG
Catalog # ANTIA318441-50
Supplier:  ANTIBODIES.COM