ContactUsLinkComponent
Anti-SARS-CoV-2 Neutralizing Antibody ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitr o quantitative determination of Anti-SARS-CoV-2 Neutralizing Antibody in serum, plasma, cell culture supernatant, cel 1 * 96 Tests
Catalog # ANTIA326874-96
Supplier:  ANTIBODIES.COM
New Product
Anti-SARS-CoV-2 Neutralizing Antibody ELISA Kit is a competitive Enzyme-Linked Immunosorbent Assay (cELISA) designed for the in vitr o quantitative determination of Anti-SARS-CoV-2 Neutralizing Antibody in serum, plasma, cell culture supernatant, cel 1 * 96 Tests
Catalog # ANTIA326874-96
Supplier:  ANTIBODIES.COM