ContactUsLinkComponent
Human Anti-SARS-CoV-2 Spike RBD (delta B.1.617.2) IgG ELISA Kit is a indirect Enzyme-Linked Immunosorbent Assay designed for the in vitro quantitative determination of human Anti-SARS-CoV-2 Spike RBD (delta B.1.617.2) IgG in serum, plasma, tissue hom 1 * 96 Tests
Catalog # ANTIA303156-96
Supplier:  ANTIBODIES.COM
New Product
Human Anti-SARS-CoV-2 Spike RBD (delta B.1.617.2) IgG ELISA Kit is a indirect Enzyme-Linked Immunosorbent Assay designed for the in vitro quantitative determination of human Anti-SARS-CoV-2 Spike RBD (delta B.1.617.2) IgG in serum, plasma, tissue hom 1 * 96 Tests
Catalog # ANTIA303156-96
Supplier:  ANTIBODIES.COM