ContactUsLinkComponent
Human MC4-R ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human MC4-R in serum, plasma, tissue homogenates and other biological fluids. 1 * 48 Tests
Catalog # ANTIA326167-48
Supplier:  Antibodies.com
New Product
Human MC4-R ELISA Kit is a sandwich Enzyme-Linked Immunosorbent Assay (sELISA) designed for the in vitro quantitative determination of human MC4-R in serum, plasma, tissue homogenates and other biological fluids. 1 * 48 Tests
Catalog # ANTIA326167-48
Supplier:  Antibodies.com