ContactUsLinkComponent
Human Anti-Respiratory Syncytial Virus IgG Antibody ELISA Kit is a 90 minute Indirect Enzyme-Linked Immunosorbent Assay designed for the in vitro qualitative determination of human Anti-Respiratory Syncytial Virus IgG antibody in serum, plasma, and n 1 * 96 Tests
Catalog # ANTIA314418-96
Supplier:  ANTIBODIES.COM
New Product
Human Anti-Respiratory Syncytial Virus IgG Antibody ELISA Kit is a 90 minute Indirect Enzyme-Linked Immunosorbent Assay designed for the in vitro qualitative determination of human Anti-Respiratory Syncytial Virus IgG antibody in serum, plasma, and n 1 * 96 Tests
Catalog # ANTIA314418-96
Supplier:  ANTIBODIES.COM