ContactUsLinkComponent
Full-length transmembrane human SLC25A4 protein expressed on the cell surface of Virus like particles. 1 * 50 µG
Catalog # ANTIA324848-50
Supplier:  Antibodies.com
New Product
Full-length transmembrane human SLC25A4 protein expressed on the cell surface of Virus like particles. 1 * 50 µG
Catalog # ANTIA324848-50
Supplier:  Antibodies.com