ContactUsLinkComponent
This Malate Dehydrogenase (NAD+/NADH) Activity Assay Kit is a quantitative assay kit for measuring Malate Dehydrogenase (NAD+/NADH) activity in animal and plant tissues, cells, bacteria, serum, plasma, and other samples 1 * 96 Tests
Catalog # ANTIA319710-96
Supplier:  ANTIBODIES.COM
New Product
This Malate Dehydrogenase (NAD+/NADH) Activity Assay Kit is a quantitative assay kit for measuring Malate Dehydrogenase (NAD+/NADH) activity in animal and plant tissues, cells, bacteria, serum, plasma, and other samples 1 * 96 Tests
Catalog # ANTIA319710-96
Supplier:  ANTIBODIES.COM